(3-Chlorophenyl)(propyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

164550-1G 164550-500MG 164550-5G 164550-25G
77465-324EA 217.33 USD
77465-324 77478-086 77488-254 77498-936
(3-Chlorophenyl)(propyl)sulfane ≥97%
(3-Chlorophenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₉H₁₁ClS
Storage Temperature: Ambient
MDL Number: MFCD26792631
CAS Number: 1713160-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR