1-Bromo-4-propylsulfanylbenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Bromophenyl)(propyl)sulphane

164548-1G 164548-500MG 164548-5G 164548-25G
77465-320EA 84.46 USD
77465-320 77478-082 77488-250 77498-932
1-Bromo-4-propylsulfanylbenzene ≥97%
1-Bromo-4-propylsulfanylbenzene

Precaución: For research use only

Formula: C₉H₁₁BrS
MW: 231.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04324355
CAS Number: 76542-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR