(2,4-Difluorophenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

164546-1G 164546-500MG 164546-5G
77465-316EA 696.28 USD
77465-316 77478-078 77488-246
(2,4-Difluorophenyl)(propyl)sulfane ≥95%
(2,4-Difluorophenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀F₂S
Storage Temperature: Ambient
MDL Number: MFCD30305999
CAS Number: 1864414-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR