2-(6-Bromo-2-fluoro-3-methylphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

16452-1G 16452-500MG 16452-5G 16452-25G
77465-312EA 331.66 USD
77465-312 77478-074 77488-238 77498-922
2-(6-Bromo-2-fluoro-3-methylphenyl)-1,3-dioxolane ≥95%
2-(6-Bromo-2-fluoro-3-methylphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₂
MW: 261.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976869
CAS Number: 2221812-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR