1,3-Dibromo-2-(methoxymethoxy)-benzene ≥95%

Supplier: AOBChem USA

Warning

16442-1G 16442-500MG 16442-5G 16442-25G
77465-308EA 188.49 USD
77465-308 77478-070 77488-234 77498-918
1,3-Dibromo-2-(methoxymethoxy)-benzene ≥95%
1,3-Dibromo-2-(methoxymethoxy)-benzene

Precaución: For research use only

Formula: C₈H₈Br₂O₂
MW: 295.956 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27998238
CAS Number: 142273-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR