1-Fluoro-2-iodo-4-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

164203-1G 164203-500MG 164203-5G 164203-25G
77465-304EA 415.09 USD
77465-304 77478-066 77488-230 77498-914
1-Fluoro-2-iodo-4-propoxybenzene ≥95%
1-Fluoro-2-iodo-4-propoxybenzene

Precaución: For research use only

Formula: C₉H₁₀FIO
MW: 280.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34850519
CAS Number: 2921859-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR