6-Methyl-[1,2,4]triazolo[4,3-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

163804-1G 163804-500MG
77465-300EA 1639.76 USD
77465-300 77478-062
6-Methyl-[1,2,4]triazolo[4,3-a]pyridine ≥97%
6-Methyl-[1,2,4]triazolo[4,3-a]pyridine

Precaución: For research use only

Formula: C₇H₇N₃
Storage Temperature: Ambient
MDL Number: MFCD09994413
CAS Number: 4919-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR