5-Bromo-7-chloro-3-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

163669-1G 163669-500MG 163669-5G
77465-296EA 639.63 USD
77465-296 77478-058 77488-224
5-Bromo-7-chloro-3-methyl-1H-indazole ≥97%
5-Bromo-7-chloro-3-methyl-1H-indazole

Precaución: For research use only

Formula: C₈H₆BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD30295613
CAS Number: 1197943-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR