(3-Bromo-2,6-dichlorophenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

163457-1G 163457-500MG 163457-5G 163457-25G
77465-292EA 415.09 USD
77465-292 77478-054 77488-220 77498-900
(3-Bromo-2,6-dichlorophenyl)(propyl)sulfane ≥95%
(3-Bromo-2,6-dichlorophenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₉H₉BrCl₂S
Storage Temperature: Ambient
MDL Number: MFCD34850005
CAS Number: 2921872-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR