(2-Bromo-4,5-difluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

162899-1G 162899-500MG 162899-5G 162899-25G
77465-284EA 116.39 USD
77465-284 77478-046 77488-212 77498-894
(2-Bromo-4,5-difluorophenyl)(methyl)sulfane ≥97%
(2-Bromo-4,5-difluorophenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₇H₅BrF₂S
MW: 239.0804 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31650117
CAS Number: 1432609-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR