3-Bromo-4-ethoxy-5-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

16236-1G 16236-5G 16236-25G
77465-276EA 60.77 USD
77465-276 77488-204 77498-892
3-Bromo-4-ethoxy-5-methoxybenzaldehyde ≥95%
3-Bromo-4-ethoxy-5-methoxybenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁BrO₃
Storage Temperature: Ambient
MDL Number: MFCD02256495
CAS Number: 90109-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR