2-Chloro-4-fluoro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16228-1G 16228-500MG 16228-5G
77465-272EA 657.14 USD
77465-272 77478-036 77488-200
2-Chloro-4-fluoro-3-methoxybenzaldehyde ≥97%
2-Chloro-4-fluoro-3-methoxybenzaldehyde

Precaución: For research use only

Formula: C₈H₆ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD24386983
CAS Number: 750586-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR