4-(4-Bromo-1-methyl-5-(trimethylsilyl)-1H-pyrazol-3-yl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

162180-1G 162180-500MG 162180-5G 162180-25G
77465-268EA 415.09 USD
77465-268 77478-032 77488-196 77498-888
4-(4-Bromo-1-methyl-5-(trimethylsilyl)-1H-pyrazol-3-yl)tetrahydro-2H-pyran-4-ol ≥95%
4-(4-Bromo-1-methyl-5-(trimethylsilyl)-1H-pyrazol-3-yl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₂H₂₁BrN₂O₂Si
MW: 333.2968 g/mol
Storage Temperature: Ambient
CAS Number: 2374145-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR