Methyl 4-bromo-3-chloro-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

162002-1G 162002-500MG 162002-5G
77465-264EA 901.25 USD
77465-264 77478-028 77488-192
Methyl 4-bromo-3-chloro-2-methoxybenzoate ≥97%
Methyl 4-bromo-3-chloro-2-methoxybenzoate

Precaución: For research use only

Formula: C₉H₈BrClO₃
Storage Temperature: Ambient
MDL Number: MFCD23707524
CAS Number: 1427377-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR