5-Amino-2,4-dichlorophenyl acetate ≥97%

Supplier: AOBChem USA

Warning

161963-1G 161963-500MG 161963-5G 161963-25G
77465-260EA 415.09 USD
77465-260 77478-024 77488-186 77498-884
5-Amino-2,4-dichlorophenyl acetate ≥97%
5-Amino-2,4-dichlorophenyl acetate

Precaución: For research use only

Formula: C₈H₇Cl₂NO₂
MW: 220.053 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34849028
CAS Number: 2969215-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR