(2,6-Dimethylphenyl)(ethyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

161567-1G 161567-500MG 161567-5G 161567-25G
77465-256EA 415.09 USD
77465-256 77478-022 77488-182 77498-876
(2,6-Dimethylphenyl)(ethyl)sulfane ≥97%
(2,6-Dimethylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₄S
Storage Temperature: Ambient
MDL Number: MFCD33404450
CAS Number: 2300138-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR