(3-Bromo-6-chloro-2-fluorophenyl)(ethyl)sulfane ≥96%

Supplier: AOBChem USA

Warning

161564-1G 161564-500MG 161564-5G 161564-25G
77465-252EA 415.09 USD
77465-252 77478-018 77488-178 77498-872
(3-Bromo-6-chloro-2-fluorophenyl)(ethyl)sulfane ≥96%
(3-Bromo-6-chloro-2-fluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₇BrClFS
Storage Temperature: Ambient
MDL Number: MFCD34848749
CAS Number: 2969475-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR