Ethyl(2-fluoro-5-methylphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

161562-1G 161562-500MG 161562-5G 161562-25G
77465-248EA 415.09 USD
77465-248 77478-014 77488-174 77498-868
Ethyl(2-fluoro-5-methylphenyl)sulfane ≥95%
Ethyl(2-fluoro-5-methylphenyl)sulfane

Precaución: For research use only

Formula: C₉H₁₁FS
Storage Temperature: Ambient
MDL Number: MFCD22484237
CAS Number: 1822765-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR