(2,5-Difluorophenyl)(ethyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

161555-1G 161555-500MG 161555-5G 161555-25G
77465-244EA 397.58 USD
77465-244 77478-010 77488-168 77498-864
(2,5-Difluorophenyl)(ethyl)sulfane ≥97%
(2,5-Difluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₈F₂S
Storage Temperature: Ambient
MDL Number: MFCD22483661
CAS Number: 1823329-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR