(3-Chloro-5-fluorophenyl)(ethyl)sulfane ≥96%

Supplier: AOBChem USA

Warning

161549-1G 161549-500MG 161549-5G 161549-25G
77465-240EA 339.9 USD
77465-240 77478-006 77488-164 77498-860
(3-Chloro-5-fluorophenyl)(ethyl)sulfane ≥96%
(3-Chloro-5-fluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₈ClFS
Storage Temperature: Ambient
MDL Number: MFCD11617718
CAS Number: 1314966-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR