4-Chloro-1-cyclopentyl-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

161026-1G 161026-500MG 161026-5G 161026-25G
77465-236EA 87.55 USD
77465-236 77478-002 77488-158 77498-852
4-Chloro-1-cyclopentyl-2-fluorobenzene ≥95%
4-Chloro-1-cyclopentyl-2-fluorobenzene

Precaución: For research use only

Formula: C₁₁H₁₂ClF
MW: 198.6643 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31429287
CAS Number: 2104744-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR