(3-Fluoro-2,6-dimethoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

16081-1G 16081-500MG 16081-5G 16081-25G
77465-228EA 258.53 USD
77465-228 77477-994 77488-148 77498-842
(3-Fluoro-2,6-dimethoxyphenyl)boronic acid ≥97%
(3-Fluoro-2,6-dimethoxyphenyl)boronic acid

Precaución: For research use only

Formula: C₈H₁₀BFO₄
Storage Temperature: Ambient
MDL Number: MFCD28365508
CAS Number: 309977-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR