1-(2,6-Difluoro-3-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160425-1G 160425-500MG 160425-5G
77465-224EA 447.02 USD
77465-224 77477-990 77488-144
1-(2,6-Difluoro-3-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2,6-Difluoro-3-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆F₂O
MW: 214.252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24310367
CAS Number: 1548111-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR