1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160416-1G 160416-500MG 160416-5G 160416-25G
77465-220EA 415.09 USD
77465-220 77477-986 77488-140 77498-836
1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆BrFOS
MW: 307.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34848024
CAS Number: 2921750-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR