1-(3,6-dibromo-2-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160401-1G 160401-500MG 160401-5G 160401-25G
77465-212EA 415.09 USD
77465-212 77477-978 77488-132 77498-828
1-(3,6-dibromo-2-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3,6-dibromo-2-fluorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₃Br₂FO
MW: 340.027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34848009
CAS Number: 2921729-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR