1-(3-Bromo-2-chloro-6-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160398-1G 160398-500MG 160398-5G 160398-25G
77465-208EA 415.09 USD
77465-208 77477-974 77488-128 77498-824
1-(3-Bromo-2-chloro-6-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-Bromo-2-chloro-6-fluorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₃BrClFO
MW: 295.576 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34848006
CAS Number: 2921834-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR