1-(5-Bromo-3-ethoxy-2-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160378-1G 160378-500MG 160378-5G 160378-25G
77465-200EA 415.09 USD
77465-200 77477-966 77488-120 77498-816
1-(5-Bromo-3-ethoxy-2-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(5-Bromo-3-ethoxy-2-fluorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₁₈BrFO₂
MW: 305.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34847987
CAS Number: 2921756-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR