1-(2-Fluoro-5-iodophenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160359-1G 160359-500MG 160359-5G 160359-25G
77465-196EA 415.09 USD
77465-196 77477-962 77488-116 77498-812
1-(2-Fluoro-5-iodophenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2-Fluoro-5-iodophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄FIO
Storage Temperature: Ambient
MDL Number: MFCD34847968
CAS Number: 2921894-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR