1-(2-Fluoro-5-methoxyphenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

160345-1G 160345-500MG 160345-5G 160345-25G
77465-192EA 415.09 USD
77465-192 77477-958 77488-112 77498-808
1-(2-Fluoro-5-methoxyphenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(2-Fluoro-5-methoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD34847955
CAS Number: 2921729-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR