1-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160342-1G 160342-500MG 160342-5G 160342-25G
77465-188EA 415.09 USD
77465-188 77477-954 77488-108 77498-804
1-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₃BrF₄O
MW: 329.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34847953
CAS Number: 2921833-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR