1-(3-Chloropyridin-4-yl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160337-1G 160337-500MG 160337-5G
77465-184EA 646.84 USD
77465-184 77477-950 77488-104
1-(3-Chloropyridin-4-yl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-Chloropyridin-4-yl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₄ClNO
MW: 199.677 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30091131
CAS Number: 1592259-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR