3-(Methoxymethoxy)pyridine ≥95%

Supplier: AOBChem USA

Warning

16018-1G 16018-500MG 16018-5G 16018-25G
77465-180EA 105.06 USD
77465-180 77477-946 77488-100 77498-798
3-(Methoxymethoxy)pyridine ≥95%
3-(Methoxymethoxy)pyridine

Precaución: For research use only

Formula: C₇H₉NO₂
MW: 139.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00965317
CAS Number: 81245-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR