(2,6-Dichloro-3-ethoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

157646-1G 157646-500MG 157646-5G 157646-25G
77465-164EA 415.09 USD
77465-164 77477-930 77488-078 77498-778
(2,6-Dichloro-3-ethoxyphenyl)(methyl)sulfane ≥95%
(2,6-Dichloro-3-ethoxyphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀Cl₂OS
Storage Temperature: Ambient
MDL Number: MFCD28360103
CAS Number: 1934630-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR