1-(Benzyloxy)-2,4-dichloro-3-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

157597-1G 157597-500MG 157597-5G 157597-25G
77465-156EA 415.09 USD
77465-156 77477-924 77488-070 77498-770
1-(Benzyloxy)-2,4-dichloro-3-iodobenzene ≥95%
1-(Benzyloxy)-2,4-dichloro-3-iodobenzene

Precaución: For research use only

Formula: C₁₃H₉Cl₂IO
Storage Temperature: Ambient
MDL Number: MFCD34846300
CAS Number: 2921885-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR