2,6-Dichloro-3-(ethylthio)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

157385-1G 157385-500MG 157385-5G 157385-25G
77465-152EA 415.09 USD
77465-152 77477-920 77488-066 77498-766
2,6-Dichloro-3-(ethylthio)benzaldehyde ≥95%
2,6-Dichloro-3-(ethylthio)benzaldehyde

Precaución: For research use only

Formula: C₉H₈Cl₂OS
MW: 235.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34846125
CAS Number: 2921834-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR