3-Acetyl-4-methoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

15685-1G 15685-500MG 15685-5G
77465-148EA 572.68 USD
77465-148 77477-916 77488-062
3-Acetyl-4-methoxyphenylboronic acid ≥95%
3-Acetyl-4-methoxyphenylboronic acid

Precaución: For research use only

Formula: C₉H₁₁BO₄
MW: 193.992 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28167274
CAS Number: 1034138-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR