8-(2-Fluoropyridin-3-yl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

156497-1G 156497-500MG 156497-5G 156497-25G
77465-144EA 415.09 USD
77465-144 77477-912 77488-058 77498-758
8-(2-Fluoropyridin-3-yl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(2-Fluoropyridin-3-yl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₃H₁₆FNO₃
MW: 253.269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34845449
CAS Number: 2921739-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR