5-Bromo-2-chloro-3-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

156372-1G 156372-500MG 156372-5G 156372-25G
77465-140EA 415.09 USD
77465-140 77477-908 77488-054 77498-754
5-Bromo-2-chloro-3-(trifluoromethyl)benzaldehyde ≥97%
5-Bromo-2-chloro-3-(trifluoromethyl)benzaldehyde

Precaución: For research use only

Formula: C₈H₃BrClF₃O
MW: 287.461 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34845404
CAS Number: 2091911-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR