2-(3-Bromo-2-chloro-6-fluorophenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

15619-1G 15619-500MG 15619-5G 15619-25G
77465-136EA 89.61 USD
77465-136 77477-904 77488-050 77498-752
2-(3-Bromo-2-chloro-6-fluorophenyl)-1,3-dioxolane ≥95%
2-(3-Bromo-2-chloro-6-fluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₇BrClFO₂
MW: 281.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976824
CAS Number: 2643367-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR