2-(2-Bromo-6-fluoro-5-methoxyphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

15607-1G 15607-500MG 15607-5G 15607-25G
77465-132EA 327.54 USD
77465-132 77477-900 77488-046 77498-748
2-(2-Bromo-6-fluoro-5-methoxyphenyl)-1,3-dioxolane ≥95%
2-(2-Bromo-6-fluoro-5-methoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₃
MW: 277.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976822
CAS Number: 2221812-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR