tert-Butyl 4-(3-fluoro-5-methylphenyl)-4-hydroxypiperidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

155833-1G 155833-500MG 155833-5G 155833-25G
77465-128EA 415.09 USD
77465-128 77477-896 77488-042 77498-744
tert-Butyl 4-(3-fluoro-5-methylphenyl)-4-hydroxypiperidine-1-carboxylate ≥95%
tert-Butyl 4-(3-fluoro-5-methylphenyl)-4-hydroxypiperidine-1-carboxylate

Precaución: For research use only

Formula: C₁₇H₂₄FNO₃
MW: 309.376 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34845022
CAS Number: 2921846-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR