tert-Butyl 4-(3-fluorophenyl)-4-hydroxypiperidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

155794-1G 155794-500MG 155794-5G 155794-25G
77465-124EA 336.81 USD
77465-124 77477-892 77488-038 77498-740
tert-Butyl 4-(3-fluorophenyl)-4-hydroxypiperidine-1-carboxylate ≥95%
tert-Butyl 4-(3-fluorophenyl)-4-hydroxypiperidine-1-carboxylate

Precaución: For research use only

Formula: C₁₆H₂₂FNO₃
MW: 295.349 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953396
CAS Number: 1191240-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR