2-Bromo-6-chloro-3-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

15561-1G 15561-500MG 15561-5G 15561-25G
77465-120EA 96.82 USD
77465-120 77477-888 77488-032 77498-734
2-Bromo-6-chloro-3-methylbenzaldehyde ≥95%
2-Bromo-6-chloro-3-methylbenzaldehyde

Precaución: For research use only

Formula: C₈H₆BrClO
MW: 233.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110255
CAS Number: 1114809-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR