2-(4-Fluorophenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

154877-1G 154877-500MG 154877-5G 154877-25G
77465-112EA 339.9 USD
77465-112 77477-880 77488-022 77498-716
2-(4-Fluorophenyl)cyclopentanol ≥95%
2-(4-Fluorophenyl)cyclopentanol

Precaución: For research use only

Formula: C₁₁H₁₃FO
MW: 180.2187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16818417
CAS Number: 1250764-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR