1-(3-Bromophenyl)heptan-1-ol ≥95%

Supplier: AOBChem USA

Warning

154603-1G 154603-500MG 154603-5G 154603-25G
77465-104EA 336.81 USD
77465-104 77477-872 77488-010 77498-700
1-(3-Bromophenyl)heptan-1-ol ≥95%
1-(3-Bromophenyl)heptan-1-ol

Precaución: For research use only

Formula: C₁₃H₁₉BrO
MW: 271.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16820773
CAS Number: 71434-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR