2-(2-Chloro-4-fluorophenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

15457-1G 15457-500MG 15457-5G 15457-25G
77465-100EA 138.02 USD
77465-100 77477-868 77488-006 77498-696
2-(2-Chloro-4-fluorophenyl)-1,3-dioxolane ≥95%
2-(2-Chloro-4-fluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD06209131
CAS Number: 773102-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR