2-Ethoxy-5-(trifluoromethoxy)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

15450-1G 15450-500MG 15450-5G 15450-25G
77465-096EA 111.24 USD
77465-096 77477-864 77488-002 77498-694
2-Ethoxy-5-(trifluoromethoxy)phenylboronic acid ≥95%
2-Ethoxy-5-(trifluoromethoxy)phenylboronic acid

Precaución: For research use only

Formula: C₉H₁₀BF₃O₄
MW: 249.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059518
CAS Number: 1310405-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR