1-Bromo-3,5-dichloro-2-(cyclopropylmethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

154019-1G 154019-500MG 154019-5G 154019-25G
77465-092EA 415.09 USD
77465-092 77477-860 77487-998 77498-692
1-Bromo-3,5-dichloro-2-(cyclopropylmethoxy)benzene ≥95%
1-Bromo-3,5-dichloro-2-(cyclopropylmethoxy)benzene

Precaución: For research use only

Formula: C₁₀H₉BrCl₂O
MW: 295.988 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34843872
CAS Number: 2921745-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR