2-Bromo-1-fluoro-3-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

153260-1G 153260-500MG 153260-5G 153260-25G
77465-088EA 293.55 USD
77465-088 77477-856 77487-994 77498-686
2-Bromo-1-fluoro-3-(methoxymethyl)benzene ≥97%
2-Bromo-1-fluoro-3-(methoxymethyl)benzene

Precaución: For research use only

Formula: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24121174
CAS Number: 1510057-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR