1,2-Difluoro-4-methoxy-5-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

152713-1G 152713-500MG 152713-5G 152713-25G
77465-080EA 83.43 USD
77465-080 77477-848 77487-986 77498-678
1,2-Difluoro-4-methoxy-5-nitrobenzene ≥97%
1,2-Difluoro-4-methoxy-5-nitrobenzene

Precaución: For research use only

Formula: C₇H₅F₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD11040445
CAS Number: 66684-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR